missing translation for 'onlineSavingsMsg'
Learn More

N-Guanylurea sulfate, 97%

Product Code. 11439406
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11439406

Brand: Thermo Scientific Alfa Aesar A19106.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-01-5 | C4H12N8O2·H2SO4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-01-5
Molecular Formula C4H12N8O2·H2SO4
MDL Number MFCD00035565
Synonym Dicyanodiamidine sulfate

Specifications

Melting Point ∼195°C (decomposition)
Quantity 50 g
Beilstein 6099371
Merck Index 14,3093
Formula Weight 302.27
Percent Purity 97%
Chemical Name or Material N-Guanylurea sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.