CAS RN 591-01-5
CAS RN 591-01-5
IUPAC Name:
bis((diaminomethylidene)urea); sulfuric acid
Synonyms:
bis(diaminomethylideneurea); sulfuric acid
Molecular Weight (g/mol):
302.27
Molecular Formula:
C4H14N8O6S
InChi Key:
IATXFPUBPMZBPH-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.NC(N)=NC(N)=O.NC(N)=NC(N)=O
1
–
4
of
4
results
1
N-Guanylurea sulfate, 97%
CAS: 591-01-5 Molecular Formula: C4H12N8O2·H2SO4 MDL Number: MFCD00035565 Synonym: Dicyanodiamidine sulfate
2
Guanylurea sulfate, 97%
CAS: 591-01-5 Molecular Formula: C4H12N8O2·H2SO4 Molecular Weight (g/mol): 302.26 MDL Number: MFCD00035565
3
Guanylurea sulfate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
4
N-Guanylurea Sulfate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.