Learn More
N-Fmoc-L-aspartic acid 4-tert-butyl ester, 98%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
L-Aspartic acid is a nonessential amino acid that is used to biosynthesize other amino acids within the human body.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Aspartic acid is a nonessential amino acid that is used to biosynthesize other amino acids within the human body.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
L-Aspartic acid is a nonessential amino acid that is used to biosynthesize other amino acids within the human body.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Spezifikation
| Melting Point | 148°C to 150°C (decomposition) |
| Quantity | 25 g |
| Beilstein | 3635671 |
| Solubility Information | Slightly soluble in water. |
| Optical Rotation | −24° (c=1 in DMF) |
| Formula Weight | 411.45 |
| Percent Purity | 98% |
| Chemical Name or Material | N-Fmoc-L-aspartic acid 4-tert-butyl ester |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur