CAS RN 71989-14-5
CAS RN 71989-14-5
IUPAC Name:
(2S)-4-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoate
Synonyms:
(2S)-4-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxobutanoate
Molecular Weight (g/mol):
410.45
Molecular Formula:
C23H24NO6
InChi Key:
FODJWPHPWBKDON-IBGZPJMESA-M
SMILES:
CC(C)(C)OC(=O)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O
1
–
1
of
1
results
1
N-Fmoc-L-aspartic acid 4-tert-butyl ester, 98%
CAS: 71989-14-5 Molecular Formula: C23H25NO6 Molecular Weight (g/mol): 411.454 MDL Number: MFCD00037131 InChI Key: FODJWPHPWBKDON-IBGZPJMESA-N Synonym: fmoc-asp otbu-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid,fmoc-asp obut,fmoc-l-aspartic acid beta-tert-butyl ester,fmoc-asp ot-bu-oh,fmoc-l-asp tbu,fmocasp otbu-oh,fmoc-asp obu-oh,fmoc-asp tbu-oh,fmoc-l-aspartic acid 4-tert-butyl ester PubChem CID: 2724635 IUPAC Name: (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid SMILES: CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13