Learn More
N-Despropyl Ropivacaine, TRC
Product Code. 30137869
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137869 1 g 1g
30136852 500 mg 500mg
30005428 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137869 Supplier Toronto Research Chemicals Supplier No. TRCD2973851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27262-40-4
Molecular Formula C14 H20 N2 O
Molecular Weight (g/mol) 232.32
Synonym 2-Piperidinecarboxamide, N-(2,6-dimethylphenyl)-, (2S)-, 2-Piperidinecarboxamide, N-(2,6-dimethylphenyl)-, (S)-, 2',6'-Pipecoloxylidide, (+)- (8CI), (2S)-N-(2,6-Dimethylphenyl)-2-piperidinecarboxamide, (S)-N-(2,6-Dimethylphenyl)-2-piperidinecarboxamide, L-Pipecolic acid 2,6-xylidide, N-Despropyl Ropivacaine
IUPAC Name (2S)-N-(2,6-dimethylphenyl)piperidine-2-carboxamide
SMILES Cc1cccc(C)c1NC(=O)[C@@H]2CCCCN2
Specifications
Quantity 1 g
InChI Formula InChI=1S/C14H20N2O/c1-10-6-5-7-11(2)13(10)16-14(17)12-8-3-4-9-15-12/h5-7,12,15H,3-4,8-9H2,1-2H3,(H,16,17)/t12-/m0/s1
Formula Weight 232.1576
Chemical Name or Material N-Despropyl Ropivacaine