accessibility menu, dialog, popup

UserName

CAS RN 27262-40-4

CH3CH3NHONH

CAS RN 27262-40-4

IUPAC Name: N-(2,6-dimethylphenyl)piperidine-2-carboxamide
Synonyms: N-(2,6-dimethylphenyl)piperidine-2-carboxamide
Molecular Weight (g/mol): 232.33
Molecular Formula: C14H20N2O
InChi Key: SILRCGDPZGQJOQ-UHFFFAOYNA-N
SMILES: CC1=CC=CC(C)=C1NC(=O)C1CCCCN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)