missing translation for 'onlineSavingsMsg'
Learn More

N-Citryl (R)-Phenylephrine (Mixture of Diastereomers), TRC

Product Code. 30135526
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135526 25 mg 25mg
30134712 50 mg 50mg
17921165 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135526 Supplier Toronto Research Chemicals Supplier No. TRCC53572025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H19NO8
Molecular Weight (g/mol) 341.31
Synonym 2-Hydroxy-2-(2-(((R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl)(methyl)amino)-2-oxoethyl)succinic Acid
IUPAC Name 2-hydroxy-2-[2-[[(2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl]-methylamino]-2-oxoethyl]butanedioic acid
SMILES OC1=CC=CC([C@@H](O)CN(C(CC(C(O)=O)(O)CC(O)=O)=O)C)=C1

Specifications

Analyte or Component Names N-Citryl (R)-Phenylephrine (Mixture of Diastereomers)
Shipping Condition Room Temperature
InChI Formula InChI=1S/C15H19NO8/c1-16(8-11(18)9-3-2-4-10(17)5-9)12(19)6-15(24,14(22)23)7-13(20)21/h2-5,11,17-18,24H,6-8H2,1H3,(H,20,21)(H,22,23)/t11-,15?/m0/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 25 mg
Chemical Name or Material N-Citryl (R)-Phenylephrine (Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.