missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cadherin Antibody (13A9), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 35 publications
Brand: Novus Biologicals NBP1-48309
This item is not returnable.
View return policy
Description
N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), Immunocytochemistry.
Specifications
| N-Cadherin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5 ug/ml, Simple Western 1:50, Flow Cytometry, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Flow (Intracellular), Immunocytochemistry | |
| cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin | |
| Mouse | |
| 140 kDa | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells | |
| 1000 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 13A9 | |
| Unconjugated | |
| P19022 | |
| CDH2 | |
| This N-Cadherin Antibody (13A9) was developed against the cytoplasmic domain of human N Cadherin [Swiss-Prot# P19022]. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction