missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-glutamic acid 5-tert-butyl ester, 99%

Product Code. 11406026
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11406026

Brand: Thermo Scientific Alfa Aesar B22322.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13726-84-6 | C14H25NO6 | 303.355 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13726-84-6
Molecular Formula C14H25NO6
Molecular Weight (g/mol) 303.355
MDL Number MFCD00038257
InChI Key YGSRAYJBEREVRB-VIFPVBQESA-N
Synonym boc-glu otbu-oh, boc-l-glu otbu-oh, s-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-l-glutamic acid 5-tert-butyl ester, n-boc-l-glutamic acid 5-tert-butyl ester, n-tert-butoxycarbonyl-l-glutamic acid gamma-tert-butyl ester, 2s-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-glu otbu, ambotzbaa1297, pubchem18923
PubChem CID 6993432
IUPAC Name (2S)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Melting Point 110°C to 112°C
Quantity 5 g
Formula Weight 303.36
Percent Purity 99%
Chemical Name or Material N-Boc-L-glutamic acid 5-tert-butyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.