CAS RN 13726-84-6
CAS RN 13726-84-6
IUPAC Name:
(2S)-5-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoate
Synonyms:
(2S)-5-(tert-butoxy)-2-[(tert-butoxycarbonyl)amino]-5-oxopentanoate
Molecular Weight (g/mol):
302.35
Molecular Formula:
C14H24NO6
InChi Key:
YGSRAYJBEREVRB-VIFPVBQESA-M
SMILES:
CC(C)(C)OC(=O)CC[C@H](NC(=O)OC(C)(C)C)C([O-])=O
1
–
1
of
1
results
1
N-Boc-L-glutamic acid 5-tert-butyl ester, 99%
CAS: 13726-84-6 Molecular Formula: C14H25NO6 Molecular Weight (g/mol): 303.355 MDL Number: MFCD00038257 InChI Key: YGSRAYJBEREVRB-VIFPVBQESA-N Synonym: boc-glu otbu-oh,boc-l-glu otbu-oh,s-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid,boc-l-glutamic acid 5-tert-butyl ester,n-boc-l-glutamic acid 5-tert-butyl ester,n-tert-butoxycarbonyl-l-glutamic acid gamma-tert-butyl ester,2s-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid,boc-glu otbu,ambotzbaa1297,pubchem18923 PubChem CID: 6993432 IUPAC Name: (2S)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid SMILES: CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C