Learn More
N-Boc-L-aspartic acid, 98%
Product Code. 11368195
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11368195
Brand: Thermo Scientific Alfa Aesar L08498.06
Request Bulk or Custom Format
CAS: 13726-67-5 | C9H15NO6 | 233.22 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13726-67-5
Molecular Formula C9H15NO6
Molecular Weight (g/mol) 233.22
MDL Number MFCD00037279
InChI Key KAJBMCZQVSQJDE-YFKPBYRVSA-N
Synonym boc-asp-oh, n-tert-butoxycarbonyl-l-aspartic acid, boc-l-aspartic acid, boc-l-asp-oh, n-boc-l-aspartic acid, boc-aspartic acid, boc-l-aspartic acid-oh, n-tert-butyloxycarbonyl-l-aspartic acid, n-t-butoxycarbonyl-l-aspatic acid, 2s-2-tert-butoxycarbonyl amino butanedioic acid
PubChem CID 99718
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O
Specifications
Melting Point 115°C
Quantity 5 g
Beilstein 1913973
Optical Rotation −5.5° (c=1 in Methanol)
Formula Weight 233.23
Percent Purity 98%
Chemical Name or Material N-Boc-L-aspartic acid