accessibility menu, dialog, popup

UserName

CAS RN 13726-67-5

CH3CH3H3COOHNHOOOHO

CAS RN 13726-67-5

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}butanedioic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]butanedioic acid
Molecular Weight (g/mol): 233.22
Molecular Formula: C9H15NO6
InChi Key: KAJBMCZQVSQJDE-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(O)=O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)