missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-aminoethyl 4-tert-Butyl-2,6-dimethyl-3-hydroxyphenylacetamide, TRC

Product Code. 30335696
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335696 5 mg 5mg
30347694 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30335696 Supplier Toronto Research Chemicals Supplier No. TRCB6200655MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-2-aminoethyl 4-tert-Butyl-2,6-dimethyl-3-hydroxyphenylacetamide
Analyte or Component Names N-Boc-2-aminoethyl 4-tert-Butyl-2,6-dimethyl-3-hydroxyphenylacetamide
Shipping Condition Room Temperature
Molecular Formula C21H34N2O4
InChI Formula InChI=1S/C21H34N2O4/c1-13-11-16(20(3,4)5)18(25)14(2)15(13)12-17(24)22-9-10-23-19(26)27-21(6,7)8/h11,25H,9-10,12H2,1-8H3,(H,22,24)(H,23,26)
Solution Type Neat
SMILES O=C(NCCNC(OC(C)(C)C)=O)CC1=C(C)C(O)=C(C(C)(C)C)C=C1C
IUPAC Name tert-butyl (2-(2-(4-(tert-butyl)-3-hydroxy-2,6-dimethylphenyl)acetamido)ethyl)carbamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 378.51
Quantity 5 mg
Formula Weight 378.25
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.