missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-leucine, 98% (dry wt.), may cont. up to ca 5% solvent, Thermo Scientific Chemicals

Product Code. 11338185
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338185 25 g 25g
11328185 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338185 Supplier Thermo Scientific Alfa Aesar Supplier No. L08471.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is employed as a reactant for peptide-based preparation of potential antitumor agents, preparation of cyclopropyl peptidomimetics and peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reactant for peptide-based preparation of potential antitumor agents, preparation of cyclopropyl peptidomimetics and peptide synthesis.

Solubility
Not miscible or difficult to mix in water. Soluble in chloroform, DMSO and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2018-66-8
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00026494
InChI Key USPFMEKVPDBMCG-LBPRGKRZSA-N
Synonym z-leu-oh, n-cbz-l-leucine, n-carbobenzoxy-l-leucine, carbobenzoxy-l-leucine, n-carbobenzyloxy-l-leucine, carbobenzoxyleucine, z-l-leucine, benzyloxycarbonyl-l-leucine, n-benzyloxycarbonyl-l-leucine, cbz-leu-oh
PubChem CID 74840
ChEBI CHEBI:28282
IUPAC Name (2S)-4-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Density 1
Flash Point >110°C (230°F)
Odor Odorless
Refractive Index 1.508
Quantity 25 g
Beilstein 1253861
Solubility Information Not miscible or difficult to mix in water. Soluble in chloroform,DMSO and methanol.
Optical Rotation −17.5° (c=2 in Ethanol)
Formula Weight 265.3
Percent Purity ≥98%
Assay (dry wt.), may cont. up to ≈5% solvent
Chemical Name or Material N-Benzyloxycarbonyl-L-leucine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.