accessibility menu, dialog, popup

UserName

H3C(S)HNOHOOOCH3

CAS RN 2018-66-8

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: USPFMEKVPDBMCG-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)