Learn More
N-Benzyloxycarbonyl-6-aminohexanoic acid, 98%
Product Code. 11332826
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11332826
Brand: Thermo Scientific Alfa Aesar L08997.06
Request Bulk or Custom Format
CAS: 1947-00-8 | C14H19NO4 | 265.309 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1947-00-8
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD00004423
InChI Key RXQDBVWDABAAHL-UHFFFAOYSA-N
Synonym z-6-aminohexanoic acid, z-epsilon-acp-oh, 6-benzyloxy carbonyl amino hexanoic acid, n-benzyloxycarbonyl-6-aminocaproic acid, cbz-, a-aminocaproic acid, n-cbz-6-aminohexanoic acid, n-benzyloxycarbonyl-6-aminohexanoic acid, n-carbobenzoxy-epsilon-aminocaproic acid, 6-phenylmethoxy carbonyl amino hexanoic acid
PubChem CID 74758
IUPAC Name 6-(phenylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NCCCCCC(=O)O
Specifications
Melting Point 53°C to 57°C
Quantity 5 g
Beilstein 2288508
Formula Weight 265.31
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-6-aminohexanoic acid