CAS RN 1947-00-8
CAS RN 1947-00-8
IUPAC Name:
6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Synonyms:
6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol):
265.31
Molecular Formula:
C14H19NO4
InChi Key:
RXQDBVWDABAAHL-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCCNC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
N-Benzyloxycarbonyl-6-aminohexanoic acid, 98%
CAS: 1947-00-8 Molecular Formula: C14H19NO4 Molecular Weight (g/mol): 265.309 MDL Number: MFCD00004423 InChI Key: RXQDBVWDABAAHL-UHFFFAOYSA-N Synonym: z-6-aminohexanoic acid,z-epsilon-acp-oh,6-benzyloxy carbonyl amino hexanoic acid,n-benzyloxycarbonyl-6-aminocaproic acid,cbz-,a-aminocaproic acid,n-cbz-6-aminohexanoic acid,n-benzyloxycarbonyl-6-aminohexanoic acid,n-carbobenzoxy-epsilon-aminocaproic acid,6-phenylmethoxy carbonyl amino hexanoic acid PubChem CID: 74758 IUPAC Name: 6-(phenylmethoxycarbonylamino)hexanoic acid SMILES: C1=CC=C(C=C1)COC(=O)NCCCCCC(=O)O