missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-4-O-benzyl Dopamine Tri-O-acetyl-3-Beta-D-Glucuronic Acid Methyl Ester, TRC
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
Specifications
Specifications
| Chemical Name or Material | N-Benzyloxycarbonyl-4-O-benzyl Dopamine Tri-O-acetyl-3-beta-D-Glucuronic Acid Methyl Ester |
| Analyte or Component Names | N-Benzyloxycarbonyl-4-O-benzyl Dopamine Tri-O-acetyl-3-beta-D-Glucuronic Acid Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C36H39NO13 |
| Synonym | 5-(2-(N-Benzyloxycarbonyl)aminoethyl)-2-(O-benzyl)hydroxyphenyl Tri-O-acetyl-β-D-glucopyranosiduronic Acid Methyl Ester |
| InChI Formula | InChI=1S/C36H39NO13/c1-22(38)46-30-31(47-23(2)39)33(48-24(3)40)35(50-32(30)34(41)43-4)49-29-19-25(15-16-28(29)44-20-26-11-7-5-8-12-26)17-18-37-36(42)45-21-27-13-9-6-10-14-27/h5-16,19,30-33,35H,17-18,20-21H2,1-4H3,(H,37,42)/t30?,31?,32-,33?,35+/m0/s1 |
| Solution Type | Neat |
| SMILES | O=C(OCC1=CC=CC=C1)NCCC2=CC(O[C@H]3[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@@H](C(OC)=O)O3)=C(OCC4=CC=CC=C4)C=C2 |
| IUPAC Name | N-Benzyloxycarbonyl-4-O-benzyl Dopamine Tri-O-acetyl-3-b-D-Glucuronic Acid Methyl Ester |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction