missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-4-O-benzyl Dopamine Tri-O-acetyl-3-Beta-D-Glucuronic Acid Methyl Ester, TRC

Product Code. 30333458
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
This item is not returnable. View return policy

Product Code. 30333458

Brand: Toronto Research Chemicals TRCB2861705MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzyloxycarbonyl-4-O-benzyl Dopamine Tri-O-acetyl-3-beta-D-Glucuronic Acid Methyl Ester
Analyte or Component Names N-Benzyloxycarbonyl-4-O-benzyl Dopamine Tri-O-acetyl-3-beta-D-Glucuronic Acid Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C36H39NO13
Synonym 5-(2-(N-Benzyloxycarbonyl)aminoethyl)-2-(O-benzyl)hydroxyphenyl Tri-O-acetyl-β-D-glucopyranosiduronic Acid Methyl Ester
InChI Formula InChI=1S/C36H39NO13/c1-22(38)46-30-31(47-23(2)39)33(48-24(3)40)35(50-32(30)34(41)43-4)49-29-19-25(15-16-28(29)44-20-26-11-7-5-8-12-26)17-18-37-36(42)45-21-27-13-9-6-10-14-27/h5-16,19,30-33,35H,17-18,20-21H2,1-4H3,(H,37,42)/t30?,31?,32-,33?,35+/m0/s1
Solution Type Neat
SMILES O=C(OCC1=CC=CC=C1)NCCC2=CC(O[C@H]3[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@@H](C(OC)=O)O3)=C(OCC4=CC=CC=C4)C=C2
IUPAC Name N-Benzyloxycarbonyl-4-O-benzyl Dopamine Tri-O-acetyl-3-b-D-Glucuronic Acid Methyl Ester
Industry Type Pharmaceutical
Molecular Weight (g/mol) 693.69
Quantity 5 mg
Formula Weight 639.24
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.