missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-4-O-benzyl Dopamine 3-Beta-D-Glucuronide Sodium Salt, TRC

Product Code. 30347796
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
This item is not returnable. View return policy

Product Code. 30347796

Brand: Toronto Research Chemicals TRCB28616525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzyloxycarbonyl-4-O-benzyl Dopamine 3-beta-D-Glucuronide Sodium Salt
Analyte or Component Names N-Benzyloxycarbonyl-4-O-benzyl Dopamine 3-beta-D-Glucuronide Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C29H30NNaO10
Synonym 5-(2-(N-Benzyloxycarbonyl)aminoethyl)-2-(O-benzyl)hydroxyphenyl β-D-Glucopyranosiduronic Acid Sodium Salt
InChI Formula InChI=1S/C29H31NO10.Na/c31-23-24(32)26(27(34)35)40-28(25(23)33)39-22-15-18(11-12-21(22)37-16-19-7-3-1-4-8-19)13-14-30-29(36)38-17-20-9-5-2-6-10-20;/h1-12,15,23-26,28,31-33H,13-14,16-17H2,(H,30,36)(H,34,35);/q;+1/p-1/t23-,24-,25+,26?,28+;/m0./s1
Solution Type Neat
SMILES O[C@H]1[C@H](O)[C@@H](O)[C@H](OC2=C(OCC3=CC=CC=C3)C=CC(CCNC(OCC4=CC=CC=C4)=O)=C2)O[C@@H]1C(O[Na])=O
IUPAC Name sodium (2S,3S,4S,5R,6S)-6-(2-(benzyloxy)-5-(2-(((benzyloxy)carbonyl)amino)ethyl)phenoxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 575.54
Quantity 25 mg
Formula Weight 575.18
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.