missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl Metaxalone, TRC

Product Code. 30346727
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
This item is not returnable. View return policy

Product Code. 30346727

Brand: Toronto Research Chemicals TRCB2850005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzyl Metaxalone
Analyte or Component Names N-Benzyl Metaxalone
Shipping Condition Room Temperature
Molecular Formula C19 H21 N O3
Synonym 3-Benzyl-5-((3,5-dimethylphenoxy)methyl)oxazolidin-2-one
InChI Formula InChI=1S/C19H21NO3/c1-14-8-15(2)10-17(9-14)22-13-18-12-20(19(21)23-18)11-16-6-4-3-5-7-16/h3-10,18H,11-13H2,1-2H3
Solution Type Neat
SMILES CC1=CC(OCC(OC2=O)CN2CC3=CC=CC=C3)=CC(C)=C1
IUPAC Name 3-benzyl-5-[(3,5-dimethylphenoxy)methyl]oxazolidin-2-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 311.37494
Quantity 5 g
Formula Weight 311.1521
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.