missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyl-4-chloropentanamide, TRC
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
Specifications
Specifications
| Chemical Name or Material | N-Benzyl-4-chloropentanamide |
| Analyte or Component Names | N-Benzyl-4-chloropentanamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C3 H8 Br O3 P |
| InChI Formula | InChI=1S/C3H8BrO3P/c4-2-1-3-8(5,6)7/h1-3H2,(H2,5,6,7) |
| Solution Type | Neat |
| SMILES | OP(=O)(O)CCCBr |
| IUPAC Name | 3-bromopropylphosphonic acid |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 202.972 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction