missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylmannosamine Kinase/GNE Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94770-0.1ml
This item is not returnable.
View return policy
Description
N-Acetylmannosamine Kinase/GNE Polyclonal antibody specifically detects N-Acetylmannosamine Kinase/GNE in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| N-Acetylmannosamine Kinase/GNE | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase, DMRV, GLCNENM, glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase, IBM2, N-acylmannosamine kinase, Uae1, UDP-GlcNAc-2-epimerase/ManAc kinase, UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase, UDP-N-acetylglucosamine-2-epimerase/N-acetylmannosamine kinase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GNE (NP_001121699.1). METYGYLQRESCFQGPHELYFKNLSKRNKQIMEKNGNNRKLRVCVATCNRADYSKLAPIMFGIKTEPEFFELDVVVLGSHLIDDYGNTYRMIEQDDFDINTRLHTIVRGEDEAAMVESVGLALVKLPDVLNRLKPDIMIVHGDRFDALAL | |
| 0.1 mL | |
| Cardiovascular Biology, Cell Biology, Signal Transduction | |
| 10020 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction