missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam N-Acetyl-D-glucosamine, 221.21 Da, >98% Purity, 25 g
Description
- Molecular Weight: 221.21 Da
- Purity: >98%
- Applications: Used to identify, differentiate and characterize N-acetyl-β-D-hexoaminidase(s)
- Composition: 100% N-Acetyl-D-glucosamine
- Storage: Store at +4°C
- Shipping: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% N-Acetyl-D-glucosamine |
| Content And Storage | Store at 4°C. |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | N-Acetyl-β-D-hexoaminidase |
| Target Molecule | N-Acetyl-β-D-hexoaminidase |
| Formulation | 100% N-Acetyl-D-glucosamine |
| Quantity | 25 g |
| For Use With (Application) | Identify, differentiate and characterize N-acetyl-β-D-hexoaminidase(s) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?