missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl Amoxicilloic Acid Disodium Salt, TRC

Product Code. 30345940
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30345940 25 mg
30335899 10 mg
30340658 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30345940 Supplier Toronto Research Chemicals Supplier No. TRCA73741025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl Amoxicilloic Acid Disodium Salt
Analyte or Component Names N-Acetyl Amoxicilloic Acid Disodium Salt
Shipping Condition Room Temperature
Molecular Formula C18H21N3Na2O7S
Synonym sodium (2R,4S)-2-((R)-((R)-2-acetamido-2-(4-hydroxyphenyl)acetamido)(carboxylato)methyl)-5,5-dimethylthiazolidine-4-carboxylate
InChI Formula InChI=1S/C18H23N3O7S.2Na/c1-8(22)19-11(9-4-6-10(23)7-5-9)14(24)20-12(16(25)26)15-21-13(17(27)28)18(2,3)29-15;;/h4-7,11-13,15,21,23H,1-3H3,(H,19,22)(H,20,24)(H,25,26)(H,27,28);;/q;2*+1/p-2/t11-,12+,13+,15-;;/m1../s1
Solution Type Neat
SMILES O=C([C@H](NC(C)=O)C1=CC=C(O)C=C1)N[C@H](C(O[Na])=O)[C@]2([H])N[C@@H](C(O[Na])=O)C(C)(C)S2
IUPAC Name disodium;(2R,4S)-2-[(R)-[[(2R)-2-acetamido-2-(4-hydroxyphenyl)acetyl]amino]-carboxylatomethyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 469.42
Quantity 25 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.