missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(9-Benzyl-6-chloro-9H-purin-2-yl)formamide, TRC
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
Specifications
Specifications
| Chemical Name or Material | N-(9-Benzyl-6-chloro-9H-purin-2-yl)formamide |
| Analyte or Component Names | N-(9-Benzyl-6-chloro-9H-purin-2-yl)formamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H12 Cl N5 O |
| InChI Formula | InChI=1S/C13H12ClN5O/c14-11-10-12(18-13(17-11)16-8-20)19(7-15-10)6-9-4-2-1-3-5-9/h1-5,7-8,11H,6H2,(H2,16,17,18,20) |
| Solution Type | Neat |
| SMILES | ClC1NC(=Nc2c1ncn2Cc3ccccc3)NC=O |
| IUPAC Name | N-(9-benzyl-6-chloro-1,6-dihydropurin-2-yl)formamide |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 289.72 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction