missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chlorophenyl)phthalimide, 98+%, Thermo Scientific™

Product Code. 11437337
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437337 5 g 5g
11447337 25 g 25g
11457337 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437337 Supplier Thermo Scientific Alfa Aesar Supplier No. A15057.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7386-21-2
Molecular Formula C14H8ClNO2
Molecular Weight (g/mol) 257.67
MDL Number MFCD00023048
InChI Key QKHKQJWODBAIMN-UHFFFAOYSA-N
Synonym n-4-chlorophenyl phthalimide, n-p-chlorophenyl phthalimide, phthalimide, n-p-chlorophenyl, 1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl, 2-4-chlorophenyl isoindoline-1,3-dione, 2-4-chlorophenyl-1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl isoindole-1,3-dione, acmc-209osn, 5-21-10-00293 beilstein handbook reference, n-4-chlorophenyl-phthalimide
PubChem CID 81867
IUPAC Name 2-(4-chlorophenyl)isoindole-1,3-dione
SMILES ClC1=CC=C(C=C1)N1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 195°C to 199°C
Quantity 5 g
Beilstein 189458
Formula Weight 257.68
Percent Purity ≥98%
Chemical Name or Material N-(4-Chlorophenyl)phthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.