missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chlorophenyl)-2,6-difluorobenzamide, TRC

Product Code. 30351406
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351406 5 mg -
30352359 25 mg 25mg
30366651 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30351406 Supplier Toronto Research Chemicals Supplier No. TRCC6099905MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122987-01-3
Molecular Formula C13H8NOF2Cl
Molecular Weight (g/mol) 267.66
IUPAC Name N-(4-chlorophenyl)-2,6-difluorobenzamide
SMILES Fc1cccc(F)c1C(=O)Nc2ccc(Cl)cc2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H8ClF2NO/c14-8-4-6-9(7-5-8)17-13(18)12-10(15)2-1-3-11(12)16/h1-7H,(H,17,18)
Formula Weight 267.0262
Chemical Name or Material N-(4-Chlorophenyl)-2,6-difluorobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.