CAS RN 122987-01-3
CAS RN 122987-01-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(4-Chlorophenyl)-2,6-difluorobenzamide, TRC
CAS: 122987-01-3 Molecular Formula: C13H8NOF2Cl Molecular Weight (g/mol): 267.66 IUPAC Name: N-(4-chlorophenyl)-2,6-difluorobenzamide SMILES: Fc1cccc(F)c1C(=O)Nc2ccc(Cl)cc2