Learn More
N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methylpyrimidin-4-yl)amino]thiazole-5-carboxamide, Mikromol™
Description
N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methylpyrimidin-4-yl)amino]thiazole-5-carboxamide
Specifications
Specifications
| Chemical Name or Material | N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methylpyrimidin-4-yl)amino]thiazole-5-carboxamide |
| CAS | 302964-08-5 |
| Analyte or Component Names | N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methylpyrimidin-4-yl)amino]thiazole-5-carboxamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H13 Cl2 N5 O S |
| Certifications/Compliance | ISO 17025 |
| Synonym | 5-Thiazolecarboxamide, N-(2-chloro-6-methylphenyl)-2-[(6-chloro-2-methyl-4-pyrimidinyl)amino]-, N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methyl-4-pyrimidinyl)amino]-5-thiazolecarboxamide, 2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide |
| InChI Formula | InChI=1S/C16H13Cl2N5OS/c1-8-4-3-5-10(17)14(8)23-15(24)11-7-19-16(25-11)22-13-6-12(18)20-9(2)21-13/h3-7H,1-2H3,(H,23,24)(H,19,20,21,22) |
| SMILES | Cc1cccc(Cl)c1NC(=O)c2cnc(Nc3cc(Cl)nc(C)n3)s2 |
| IUPAC Name | N-(2-chloro-6-methylphenyl)-2-[(6-chloro-2-methylpyrimidin-4-yl)amino]-1,3-thiazole-5-carboxamide |
| Show More |