Learn More
N-[(1S)-2-Amino-2-oxo-1-(phenylmethyl)ethyl]-2-pyrazinecarboxamide, TRC
Product Code. 30136800
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136800 500 mg 500mg
30133078 10 mg 10mg
30008121 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136800 Supplier Toronto Research Chemicals Supplier No. TRCA622540500MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 289472-80-6
Molecular Formula C14 H14 N4 O2
Molecular Weight (g/mol) 270.29
Synonym 2-Pyrazinecarboxamide, N-[(1S)-2-amino-2-oxo-1-(phenylmethyl)ethyl]-, N-[(1S)-2-Amino-1-benzyl-2-oxo-ethyl]pyrazine-2-carboxamide
IUPAC Name N-[(2S)-1-amino-1-oxo-3-phenylpropan-2-yl]pyrazine-2-carboxamide
SMILES NC(=O)[C@H](Cc1ccccc1)NC(=O)c2cnccn2
Specifications
Analyte or Component Names N-[(1S)-2-Amino-1-benzyl-2-oxoethyl]pyrazine-2-carboxamide
Shipping Condition Room Temperature
InChI Formula InChI=1S/C14H14N4O2/c15-13(19)11(8-10-4-2-1-3-5-10)18-14(20)12-9-16-6-7-17-12/h1-7,9,11H,8H2,(H2,15,19)(H,18,20)/t11-/m0/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 500 mg
Formula Weight 270.1117
Chemical Name or Material N-[(1S)-2-Amino-1-benzyl-2-oxoethyl]pyrazine-2-carboxamide