Learn More
N-[(1S)-2-Amino-1-benzyl-2-oxoethyl]pyrazine-2-carboxamide, Mikromol™
Description
N-[(1S)-2-Amino-1-benzyl-2-oxoethyl]pyrazine-2-carboxamide, Mikromol
Specifications
Specifications
| Chemical Name or Material | N-[(1S)-2-Amino-1-benzyl-2-oxoethyl]pyrazine-2-carboxamide |
| CAS | 289472-80-6 |
| Analyte or Component Names | N-[(1S)-2-Amino-1-benzyl-2-oxoethyl]pyrazine-2-carboxamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H14 N4 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Pyrazinecarboxamide, N-[(1S)-2-amino-2-oxo-1-(phenylmethyl)ethyl]-, N-[(1S)-2-Amino-1-benzyl-2-oxo-ethyl]pyrazine-2-carboxamide |
| InChI Formula | InChI=1S/C14H14N4O2/c15-13(19)11(8-10-4-2-1-3-5-10)18-14(20)12-9-16-6-7-17-12/h1-7,9,11H,8H2,(H2,15,19)(H,18,20)/t11-/m0/s1 |
| Solution Type | Neat |
| SMILES | NC(=O)[C@H](Cc1ccccc1)NC(=O)c2cnccn2 |
| Show More |