missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYST3, Mouse anti-Human, Clone: 4D8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00007994-M07.100ug
This item is not returnable.
View return policy
Description
Sequence: ALPKPRNHGKLDNKQNVDWNKLIKRAVEGLAESGGSTLKSIERFLKGQKDVSALFGGSAASGFHQQLRLAIKRAIGHGRLLKDGPLYRLNTKATNVDGKSpecifications
| MYST3 | |
| Monoclonal | |
| Unconjugated | |
| MYST histone acetyltransferase (monocytic leukemia) 3 | |
| KAT6A/MGC167033/MOZ/RUNXBP2/ZNF220 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4D8 | |
| In 1x PBS, pH 7.4 | |
| NM_006766 | |
| MYST3 | |
| MYST3 (NP_006757, 81 a.a. ∼ 179 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7994 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction