missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MYRIP Polyclonal antibody specifically detects MYRIP in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MYRIP |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | DKFZp586F1018, exophilin-8, FLJ44025, MGC130034, myosin VIIA and Rab interacting protein, Myosin-VIIa- and Rab-interacting protein, MyRIP, rab effector MyRIP, SLAC2-C, SLAC2CMGC130035, Slp homolog lacking C2 domains c, Slp homologue lacking C2 domains, synaptotagmin-like protein homologue lacking C2 domains-c |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 551-770 of human MYRIP (NP_001271354.1). VSLCNISTEVLKVINATEELIAGSTGPWESPQVPPDRQKGMFPRGTDQVRLDEQLTSLEENVYLAAGTVYGLETQLTELEDAARCIHSGTDETHLADLEDQVATAAAQVHHAELQISDIESRISALTIAGLNIAPCVRFTRRRDQKQRTQVQTIDTSRQQRRKLPAPPVKAEKIETSSVTTIKTFNHNFILQGSSTNRTKERKGTTKDLMEPALESAVMY |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?