missing translation for 'onlineSavingsMsg'
Learn More
Learn More
myosin X Antibody (CL9025) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88927C
This item is not returnable.
View return policy
Description
myosin X Monoclonal antibody specifically detects myosin X in Human samples. It is validated for Western Blot
Specifications
| myosin X | |
| Monoclonal | |
| DyLight 650 | |
| FLJ10639, FLJ21066, FLJ22268, KIAA0799FLJ43256, MGC131988, myosin X, myosin-X, Unconventional myosin-10 | |
| This antibody was developed using a synthetic peptide derived from Q9HD67, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 4651 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL9025 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction