missing translation for 'onlineSavingsMsg'
Learn More

Myosin Light Chain 2 Antibody (7C9), Novus Biologicals™

Product Code. 18206390 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18206390 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18206390 Supplier Novus Biologicals Supplier No. NBP128871

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myosin Light Chain 2 Monoclonal specifically detects Myosin Light Chain 2 in Human, Rat samples. It is validated for Western Blot, ELISA, Single Cell Western.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin Light Chain 2
Applications Western Blot, ELISA, Western Blot
Classification Monoclonal
Clone 7C9
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000, Single Cell Western 1:10
Formulation Ascites with 0.03% Sodium Azide
Gene Alias cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2
Gene Symbols MYL2
Host Species Mouse
Immunogen Purified recombinant fragment of MYL2 expressed in E. Coli.
Molecular Weight of Antigen 19 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4633
Test Specificity Specific for MYL2.
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.