missing translation for 'onlineSavingsMsg'
Learn More

Myosin Heavy Chain Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 57 publications

Brand:  R&D Systems MAB4470-SP

Product Code. 18712028

  • ONLINE EXCLUSIVE PRICE
    245.00€
    231.00€ / 25µg
    Save 14.00€
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Myosin Heavy Chain Monoclonal specifically detects Myosin Heavy Chain in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Myosin Heavy Chain
Monoclonal
LYOPH
Western Blot, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL
DFNA4, FLJ13881, FLJ43092, KIAA2034DKFZp667A1311, MHC16, MYH14 variant protein, MYH17, MYHC, myosin, Myosin heavy chain 14, Myosin heavy chain, non-muscle IIc, myosin, heavy chain 14, myosin, heavy chain 14, non-muscle, myosin, heavy polypeptide 14, myosin-14, NMHC II-C, NMHC-II-C, nonmuscle myosin heavy chain II-C, Non-muscle myosin heavy chain IIc
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
79784
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b
Western Blot, Immunohistochemistry, Immunocytochemistry
MF20
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
MYH14
Chicken pectoralis-derived Myosin
25 μg
Primary
Detects Myosin Heavy Chain in human, mouse, rat and other mammalian, avian, and amphibian species.
Multi-species
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only