missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 11 Antibody (CL14619) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26875MFV450
This item is not returnable.
View return policy
Description
Myosin heavy chain 11 Monoclonal antibody specifically detects Myosin heavy chain 11 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Myosin heavy chain 11 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain | |
| This antibody was developed using a recombinant protein derived from P35749, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Hypoxia, Stem Cell Markers | |
| 4629 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot | |
| CL14619 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction