missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MYO19 Monoclonal antibody specifically detects MYO19 in Human,Monkey,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | MYO19 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Formulation | PBS (pH 7.3), 50% glycerol, 0.05% BSA |
| Gene Alias | FLJ22865, MYOHD1, myosin head domain containing 1, Myosin head domain-containing protein 1, myosin XIX, myosin-XIX |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 450-650 of human MYO19 (Q96H55).,, Sequence:, QQEEYAVEGLEWSFINYQDNQPCLDLIEGSPISICSLINEECRLNRPSSAAQLQTRIETALAGSPCLGHNKLSREPSFIVVHYAGPVRYHTAGLVEKNKDPIPPELTRLLQQSQDPLLMGLFPTNPKEKTQEEPPGQSRAPVLTVVSKFKASLEQLLQVLHSTTPHYIRCIKPNSQGQAQTFLQEEVLSQLEACGLVETIH |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?