missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYO19 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33243-100ul
This item is not returnable.
View return policy
Description
MYO19 Monoclonal antibody specifically detects MYO19 in Human,Monkey,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| MYO19 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| FLJ22865, MYOHD1, myosin head domain containing 1, Myosin head domain-containing protein 1, myosin XIX, myosin-XIX | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-650 of human MYO19 (Q96H55).,, Sequence:, QQEEYAVEGLEWSFINYQDNQPCLDLIEGSPISICSLINEECRLNRPSSAAQLQTRIETALAGSPCLGHNKLSREPSFIVVHYAGPVRYHTAGLVEKNKDPIPPELTRLLQQSQDPLLMGLFPTNPKEKTQEEPPGQSRAPVLTVVSKFKASLEQLLQVLHSTTPHYIRCIKPNSQGQAQTFLQEEVLSQLEACGLVETIH | |
| 100 μL | |
| Primary | |
| Human, Monkey, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 80179 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction