missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
| Antigen | MYL7 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MYL7 Polyclonal antibody specifically detects MYL7 in Mouse samples. It is validated for Western BlotSpecifications
| MYL7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 58498 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| atrial isoform, light polypeptide 7, regulatory, MLC-2a, MYL2A, MYLC2AMLC2a, myosin, light chain 7, regulatory | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-175 of human MYL7 (NP_067046.1). MASRKAGTRGKVAATKQAQRGSSNVFSMFEQAQIQEFKEAFSCIDQNRDGIICKADLRETYSQLGKVSVPEEELDAMLQEGKGPINFTVFLTLFGEKLNGTDPEEAILSAFRMFDPSGKGVVNKDEFKQLLLTQADKFSPAEVEQMFALTPMDLAGNIDYKSLCYIITHGDEKEE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title