missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MYH7 Antibody (MYH7/9183), Novus Biologicals™

Mouse Monoclonal Antibody

232.00€ - 462.00€

Specifications

Antigen MYH7
Clone MYH7/9183
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043618
View Documents
Novus Biologicals™
NBP3-26826-100ug
100 μg
462.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043502
View Documents
Novus Biologicals™
NBP3-26826-20ug
20 μg
232.00€
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MYH7 Monoclonal antibody specifically detects MYH7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

MYH7
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cell Biology, Cytoskeleton Markers, Signal Transduction
10 mM PBS with 0.05% BSA
4625
IgG2b κ
Protein A or G purified
MYH7/9183
Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Monoclonal
Purified
RUO
Human
cardiac muscle beta isoform, CMD1SMGC138376, CMH1, DKFZp451F047, MGC138378, MPD1, MYHCB, MyHC-beta, myhc-slow, myopathy, distal 1, myosin heavy chain (AA 1-96), myosin, heavy chain 7, cardiac muscle, beta, myosin, heavy polypeptide 7, cardiac muscle, beta, myosin-7, rhabdomyosarcoma antigen MU-RMS-40.7A, SPMD, SPMM
Recombinant fragment (around aa1150-1350) of human MYH7 protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.