missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
214.00€ - 506.00€
Specifications
| Antigen | MYH7 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18691972
|
Novus Biologicals
NBP2-94079-0.02ml |
0.02 mL |
214.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600912
|
Novus Biologicals
NBP2-94079-0.1ml |
0.1 mL |
506.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYH7 Polyclonal antibody specifically detects MYH7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| MYH7 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 4625 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cardiac muscle beta isoform, CMD1SMGC138376, CMH1, DKFZp451F047, MGC138378, MPD1, MYHCB, MyHC-beta, myhc-slow, myopathy, distal 1, myosin heavy chain (AA 1-96), myosin, heavy chain 7, cardiac muscle, beta, myosin, heavy polypeptide 7, cardiac muscle, beta, myosin-7, rhabdomyosarcoma antigen MU-RMS-40.7A, SPMD, SPMM | |
| A synthetic peptide corresponding to a sequence within amino acids 1700-1800 of human MYH7 (NP_000248.2). KKMDADLSQLQTEVEEA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title