Learn More
Myeloperoxidase/MPO Antibody (CL15002) [Alexa Fluor™ 647], Novus Biologicals™
Product Code. 30301051 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30301051 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30301051 Supplier Novus Biologicals Supplier No. NBP326847AF647
Mouse Monoclonal Antibody
Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen Myeloperoxidase/MPO
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone CL15002
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias EC 1.11.1, EC 1.11.1.7, myeloperoxidase
Host Species Mouse
Immunogen This antibody was developed using a synthetic peptide derived from P05164, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4353
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1