missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myelin Protein Zero Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-94909
This item is not returnable.
View return policy
Description
Myelin Protein Zero Polyclonal specifically detects Myelin Protein Zero in Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Myelin Protein Zero | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Charcot-Marie-Tooth neuropathy 1B, CHM, CMT1, CMT1B, CMT2I, CMT2J, CMT4E, CMTDI3, HMSNIB, MPP, Myelin peripheral protein, myelin protein P0, myelin protein zeroDSS, P0 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The presence of this PNS protein in Rat Spinal Cord is likely due to contamination with spinal roots during extraction from the animal. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_000521.1 | |
| MPZ | |
| Peptide with sequence C-DHSRSTKAVSEK corresponding to internal region (near C-Terminus) according to NP_000521.1. | |
| 0.1 mg | |
| Neuronal Cell Markers, Oligodendrocyte Cell Markers | |
| 4359 | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction