missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myelin PLP Antibody (plpc 1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-960-0.1mg
This item is not returnable.
View return policy
Description
Myelin PLP Monoclonal specifically detects Myelin PLP in Bovine, Mammal samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Myelin PLP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| HLD1, lipophilin, major myelin proteolipid protein, MMPL, myelin proteolipid protein, PLP, PLP/DM20, PMD, proteolipid protein 1, spastic paraplegia 2, uncomplicated, SPG2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5354 | |
| Bovine | |
| Purified |
| Western Blot | |
| plpc 1 | |
| Unconjugated | |
| P04116 | |
| PLP1 | |
| Synthetic peptide: C-GRGTKF, corresponding to amino acids 272-277 of Myelin PLP. | |
| 0.1 mg | |
| Primary | |
| This recognizes myelin proteolipid protein. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction