missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MyD88 specifically detects MyD88 in Human, Mouse, Rat, Bovine, Xenopus, Yeast, Zebrafish samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, In vitro assay, In vivo assay, Functional (Inhibition), Binding Inhibition, Block/Neutralize.
Specifications
Specifications
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, In vitro Assay, In vivo Assay, Functional Inhibition Assay, Binding Inhibition Assay, Blocking Assay, Neutralization |
| Host Species | Human, Mouse, Rat, Xenopus, Zebrafish |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Content And Storage | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
| Product Type | Reactivity: Hu, Mu, Rt, Xp, Ze |
| Product Line | Novus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?