missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYCT1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05580-20ul
This item is not returnable.
View return policy
Description
MYCT1 Polyclonal antibody specifically detects MYCT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| MYCT1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| FLJ21269, MGC156309, MGC156310, MTLC, MTLCmyc target in myeloid cells 1, MTMC1, myc target 1, Myc target in myeloid cells protein 1, myc target protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 99-235 of human MYCT1 (NP_079383.2). ASAPISQWSSSRRSRSSYTHGLNRTGFYRHSGCERRSNLSLASLTFQRQASLEQANSFPRKSSFRASTFHPFLQCPPLPVETESQLVTLPSSNISPTISTSHSLSRPDYWSSNSLRVGLSTPPPPAYESIIKAFPDS | |
| 20 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 80177 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction