missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolate Sodium, Mikromol™

Product Code. 30049285
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049285 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049285 Supplier Mikromol Supplier No. MM3657.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Mycophenolate Sodium
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mycophenolate Sodium
CAS 37415-62-6
Analyte or Component Names Mycophenolate Sodium
Shipping Condition Room Temperature
Molecular Formula C17 H19 O6 . Na
Certifications/Compliance ISO 17025
Synonym Mycophenolate Sodium, (4E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic acid sodium salt (1:1), 4-Hexenoic acid, 6-(1,3-dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-, monosodium salt, (4E)- (9CI), 4-Hexenoic acid, 6-(1,3-dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-, monosodium salt, (E)-, EC-MPS, ERL 080, ERL 080A, Femulan, Mycophenolate sodium, Mycophenolic acid monosodium salt, Mycophenolic monosodium salt, Myfortic, Sodium mycophenolate
InChI Formula InChI=1S/C17H20O6.Na/c1-9(5-7-13(18)19)4-6-11-15(20)14-12(8-23-17(14)21)10(2)16(11)22-3;/h4,20H,5-8H2,1-3H3,(H,18,19);/q;+1/p-1/b9-4+;
SMILES [Na+].COc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)\CCC(=O)[O-]
IUPAC Name sodium;(E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
Molecular Weight (g/mol) 342.32
Quantity 250 mg
Formula Weight 342.1079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.