missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYCBP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27951-1-AP-150UL
This item is not returnable.
View return policy
Description
SNX33 (sorting nexin-33), also known as SH3PX3, SH3PXD3C or SNX30, is a 574 amino acid protein that interacts with ADAM15 and FAS-L. Belonging to the sorting nexin family, SNX33 contains one BAR domain, one PX (phox homology) domain and one SH3 domain. The gene that encodes SNX33 consists of over 14,000 bases and maps to human chromosome 15q24.2. Housing approximately 106 million base pairs and encoding more than 700 genes, chromosome 15 makes up about 3% of the human genome. Angelman and Prader-Willi syndromes are associated with loss of function or deletion of genes in the 15q11-q13 region.Specifications
| MYCBP2 | |
| Polyclonal | |
| Unconjugated | |
| MYCBP2 | |
| DKFZp686M08244, FLJ10106, FLJ13826, FLJ21597, FLJ21646, KIAA0916, MYC binding protein 2, MYCBP2, PAM, Pam/highwire/rpm 1 protein, Protein associated with Myc | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23077 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.67 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75592 | |
| MYCBP2 | |
| MYCBP2 Fusion Protein Ag27228 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction